Learn More
Analytical Sales & Services Select-A-Vial™, 350µL Complete System
Catalog No. 5000805117
Encompass
Select-A-Vial™, 350µL Complete System\n• 96 Position\n• Racked 350µL Clear Conical Glass Inserts\n• Includes Rack and Base Tray
Catalog No. 50-008-05117 Supplier Analytical Sales & Services Supplier No. 27350
Select-A-Vial™, 350µL Complete System\n• 96 Position\n• Racked 350µL Clear Conical Glass Inserts\n• Includes Rack and Base Tray