Learn More
Analytical Sales & Services Select-A-Vial™, 500µL Silanized Complete System
Catalog No. 5000807069
Encompass
96 Position Select A Vial - Complete System Racked 500µL Silanized Clear Conical Glass Inserts in Rack and Base Tray
Catalog No. 50-008-07069 Supplier Analytical Sales & Services Supplier No. 27500S
96 Position Select A Vial - Complete System Racked 500µL Silanized Clear Conical Glass Inserts in Rack and Base Tray