Learn More
StatLab™ Select Acid Rinse
GSA_VASDP
Catalog No. STLSL404
Change view
Click to view available options
Quantity:
4 x 1 gal.
Catalog No. Quantity
STLSL404 4 x 1 gal.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL404 Supplier StatLab™ Supplier No. SL404
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Produces crisp nuclear chromatin detail and eliminates undesirable background staining.
  • Utilizes a buffer system to prevent pH fluctuation
  • Premixed and ready to use
  • Non-hazardous
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 1 gal.
Product Type Acid Rinse