Learn More
Leica Biosystems SELECT CHEK EOSIN 500ML 4/CS
Catalog No. NC0320971
Encompass_Preferred
SelecTech is a hematoxylin and eosin staining system consisting of Hematoxylin 560, Blue Buffer 8, Define, Alcoholic Eosin Y 515 and Eosin Trichrome 515.SelecTech standardizes the reagents and stains used in the hematoxylin and eosin staining procedure. The built-in quality assurance parameters assure consistent results. The reagents and stains in SelecTech are designed to complement each other to produce a crisp, well delineated hematoxylin and eosin stain.
Catalog No. NC0320971 Supplier Leica Biosystems Supplier No. 3801619
SelecTech is a hematoxylin and eosin staining system consisting of Hematoxylin 560, Blue Buffer 8, Define, Alcoholic Eosin Y 515 and Eosin Trichrome 515.SelecTech standardizes the reagents and stains used in the hematoxylin and eosin staining procedure. The built-in quality assurance parameters assure consistent results. The reagents and stains in SelecTech are designed to complement each other to produce a crisp, well delineated hematoxylin and eosin stain.