Learn More
StatLab™ Select Eosin Stain
GSA_VASDP
Catalog No. STLSL402
Change view
Click to view available options
Quantity:
4 x 16 oz.
Catalog No. Quantity
STLSL402 4 x 16 oz.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL402 Supplier StatLab™ Supplier No. SL402
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Alcoholic and acidified formulation producing a cytoplasmic stain with three distinct color variations in a short amount of staining time.
  • Produces a brilliant cytoplasmic stain
  • Proprietary formulation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 16 oz.
Product Type Eosin Stain