Learn More
Echelon Biosciences Research Labs Select-HA HiLadder
SDP
Catalog No. NC1744599
Encompass
Select-HA HiLadder is a hyaluronic acid (HA) preparation of uniform and narrow size distribution prepared by in vitro synthesis using recombinant Pasteurella multocida hyaluronan synthase. Each Select-HA Ladder™ contains 5 Select-HA™ molecular mass markers. This item is non returnable
This item is not returnable. View return policy
Catalog No. NC1744599 Supplier Echelon Biosciences Research Labs Supplier No. HYAHILAD201VIAL
Select-HA HiLadder is a hyaluronic acid (HA) preparation of uniform and narrow size distribution prepared by in vitro synthesis using recombinant Pasteurella multocida hyaluronan synthase. Each Select-HA Ladder™ contains 5 Select-HA™ molecular mass markers. This item is non returnable