Learn More
Echelon Biosciences Research Labs Select-HA LoLadder
SDP
Supplier:  Echelon Biosciences Research Labs HYALOLAD1VIAL
Encompass
Select-HA LoLadder is a hyaluronic acid (HA) preparation of uniform and narrow size distribution prepared by in vitro synthesis using recombinant Pasteurella multocida hyaluronan synthase1. Each Select-HA Ladder™ contains 5 Select-HA™ molecular mass markers. This item is non returnable
Catalog No. NC1744600