Learn More
StatLab™ Select Hematoxylin Stain
GSA_VASDP
Catalog No. STLSL401
Change view
Click to view available options
Quantity:
4 x 16 oz.
Catalog No. Quantity
STLSL401 4 x 16 oz.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL401 Supplier StatLab™ Supplier No. SL401
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Possesses the rapid staining qualities of Gill Hematoxylins along with the nuclear chromatin delineation of a Harris stain.
  • Does not require daily filtering
  • Has no affinity for background or mucin staining
  • For H&E staining, IHC counterstaining, frozen section staining'
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 16 oz.
Product Type Hematoxylin