Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Select Leather Double Palm Gloves

Catalog No. 19013483 Shop All Fisherbrand Products
Change view
Click to view available options
Glove Cuff Style:
Gauntlet
Safety
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glove Cuff Style
19-013483 Gauntlet
19-013-475 Safety
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-013483
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Constructed from select-grade shoulder split cowhide leather.

  • Gloves feature either a 2 in. rubberized safety cuff or 4 in. rubberized gauntlet cuff
  • Leather covers palm, index finger, and thumb
  • Gunn-cut construction provides longer life
  • Used for construction, metal-work, general maintenance, warehousing, and other medium- to light-duty applications

Specifications

Cuff Material Rubberized
Color Gray, Green, Pink
Grade Select
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type General Purpose Gloves
Size Large
Style Gunn
Glove Cuff Color Green with Pink Stripes
Glove Cuff Style Gauntlet
Hem Color Brown
Liner Material Cotton
Palm Type Double
Length (English) Cuff 4 in.
Length (Metric) Cuff 10.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.