Learn More
Waters Corp Performance Maintenance Kit, SELECT SERIES Cyclic IMS, Annual
Catalog No. 502391422 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit for the SELECT SERIES Cyclic IMS Mass Spectrometer contains all the necessary components to maintain the instrument’s peak performance on an annual basis.
  • Keeps the SELECT SERIES Cyclic IMS Mass Spectrometer operating at peak performance annually.
  • Includes source components, air filters, and lock spray reference probe.
  • Does not include a probe assembly; ordering a separate appropriate probe assembly is highly recommended for peak performance.
Catalog No. 50-239-1422 Supplier Waters Corp Supplier No. 201000327
This Performance Maintenance Kit for the SELECT SERIES Cyclic IMS Mass Spectrometer contains all the necessary components to maintain the instrument’s peak performance on an annual basis.
  • Keeps the SELECT SERIES Cyclic IMS Mass Spectrometer operating at peak performance annually.
  • Includes source components, air filters, and lock spray reference probe.
  • Does not include a probe assembly; ordering a separate appropriate probe assembly is highly recommended for peak performance.