Learn More
Norell Inc 5mm, 500MHz 7" SELECT SERIES NMR TUBE, 5 Tubes per pack
SDP
Catalog No. NC2851585
Encompass_Preferred
Select Series NMR Tubes are ideal for cold refrigeration storage, work with cryo-probes and, variable temperature studies. Key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or under any other application where large temperature variations are required in the experiment.
Catalog No. NC2851585 Supplier Norell Inc Supplier No. S55007
Select Series NMR Tubes are ideal for cold refrigeration storage, work with cryo-probes and, variable temperature studies. Key properties that make this glass type desirable for NMR are its high degree of thermal shock resistance and low expansion coefficient. This allows for a greater margin of safety from breakage when used in variable temperature applications and freeze/thaw cycling, or under any other application where large temperature variations are required in the experiment.