Learn More
MSC SPI Inspection Surface Plate Cleaner, 5 gal., Pail, 1 ea
Catalog No. 502867008
Encompass
This cleaner is designed for cleaning, degreasing, and caring for granite inspection surface plates. It comes in a 5 gal. pail for convenient use.
  • Packaged in a pail
  • Volume of 5 gal.
  • Designed for cleaning, degreasing, and care of granite surfaces
Catalog No. 50-286-7008 Supplier MSC Supplier No. 01310523
This cleaner is designed for cleaning, degreasing, and caring for granite inspection surface plates. It comes in a 5 gal. pail for convenient use.
  • Packaged in a pail
  • Volume of 5 gal.
  • Designed for cleaning, degreasing, and care of granite surfaces