Learn More
Zymo Research Corporation Select-A-Size Binding Buffer, 15 ml
SDP
Catalog No. 501251444
Encompass_Preferred
Select-A-Size Binding Buffer (15 ml) for use with Select-A-Size DNA Clean & Concentrator™ kits. Assists in binding nucleic acid to Zymo-Spin™ columns for size selection and purification.
Catalog No. 50-125-1444 Supplier Zymo Research Corporation Supplier No. D4080115
Select-A-Size Binding Buffer (15 ml) for use with Select-A-Size DNA Clean & Concentrator™ kits. Assists in binding nucleic acid to Zymo-Spin™ columns for size selection and purification.