Learn More
Zymo Research Corporation Select-a-Size DNA MagBead Kit
SDP
Catalog No. NC1481263
Encompass_Preferred
This simple workflow allows for specific cutoffs thatcan be modified to suit reaction clean-up, left-sided, right-sided or even double-sided size selection. The Select-a-Size MagBead Buffer selectively binds fragments based on the volume added relative to the sample. Simply choose a desired cutoff to bind the target of interest onto the beads and remove species outside of this range. The desired DNA is easily eluted from the beads following a rapid wash regimen.
Catalog No. NC1481263 Supplier Zymo Research Corporation Supplier No. D4085
This simple workflow allows for specific cutoffs thatcan be modified to suit reaction clean-up, left-sided, right-sided or even double-sided size selection. The Select-a-Size MagBead Buffer selectively binds fragments based on the volume added relative to the sample. Simply choose a desired cutoff to bind the target of interest onto the beads and remove species outside of this range. The desired DNA is easily eluted from the beads following a rapid wash regimen.