Learn More
Cayman Chemical SelectInCD62E EXrAClula 100ug
SDP
Catalog No. 502910202
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged E-selectin expressed in HEK293 cells • Amino acids: 22-556 • MW: 60 kDa
Catalog No. 50-291-0202 Supplier Cayman Chemical Supplier No. 35725100 UG
Source: Recombinant human C-terminal His-tagged E-selectin expressed in HEK293 cells • Amino acids: 22-556 • MW: 60 kDa