Learn More
Teknova Selective Growth Broth SD minus Trp. 1000ml Sterile 2 pack.
SDP
Catalog No. 502387422
Encompass
Carbon: 2.0% Cerelose Anhydrous Dextrose (Glucose),Nitrogen: 0.5% Ammonium Sulfate
Catalog No. 50-238-7422 Supplier Teknova Supplier No. S10402PK
Carbon: 2.0% Cerelose Anhydrous Dextrose (Glucose),Nitrogen: 0.5% Ammonium Sulfate