Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Bis(2-ethylhexyl) adipate, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111012336 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2336 5 mL
11-101-2335 1 mL
11-101-2334 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-2336 Supplier MilliporeSigma Supelco Supplier No. 021385ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00009496
Synonym Adipic acid di(2-ethylhexyl) ester; DOA

Specifications

Melting Point <-70°C
Density 0.925 g/mL (at 20°C)
CAS 103-23-1
Boiling Point 175°C (2 mmHg)
Flash Point 196°C - Closed Cup
Linear Formula [-CH2 CH2 CO2 CH2 CH(C2H5)(CH2)3 CH3]2
Quantity 5 mL
Formula Weight 370.57
Percent Purity ≥99.0% (GC); ≥99.0%
Grade Selectophore
Product Line Selectophore
Show More Show Less
RTECSNumber AU9700000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.