Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Calcium ionophore I, Function Tested, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014768 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4768 50 mg
11-101-4767 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4768 Supplier MilliporeSigma Supelco Supplier No. 2119250MGF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00042943
Synonym (-)-(R,R)-N,N′-Bis-[11-(ethoxycarbonyl)undecyl]-N,N′,4,5-tetramethyl-3,6-dioxaoctane-diamide; Diethyl N,N′-[(4 R,5 R)-4,5-dimethyl-1,8-dioxo-3,6-dioxaoctamethylene]bis(12-methylaminododecanoate); ETH 1001

Specifications

CAS 58801-34-6
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C38H72N2O8
Quantity 50 mg
Formula Weight 684.99
Percent Purity ≥99.0% (HPLC); ≥99.0%
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.