Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Calcium ionophore III, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111014459 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4459 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4459 Supplier MilliporeSigma Supelco Supplier No. 211865MGF

missing translation for 'validMainframeMsg'

Calcium ionophore III (Calimycin;CA) is a divalent cation ionophore. It has ability to transport Ca2+ ions across membranes along a concentration gradient.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00151202
Synonym Calcium Ionophore A23187; Antibiotic A 23187; Calimycin

Specifications

CAS 52665-69-7
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C29H37N3O6
Quantity 5 mg
Formula Weight 523.62
Grade Selectophore
Product Line Selectophore
RTECSNumber DM4676000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.