Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Carbonate ionophore VII, MilliporeSigma™ Supelco™

Catalog No. 111013574 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3574 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3574 Supplier MilliporeSigma Supelco Supplier No. 9320650MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbonate ionophore VII is an ionophore mostly used in the preparation of ion selective electrodes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD21363129
Synonym N,N-Dioctyl-3 alpha,12 alpha-bis(4-trifluoroacetylbenzoyloxy)-5 beta-cholan-24-amide

Specifications

CAS 222310-82-9
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C58H79F6 NO7
Quantity 50 mg
Formula Weight 1016.24
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.