Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Copper(II) ionophore I, MilliporeSigma™ Supelco™

Catalog No. 111012847 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2847 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2847 Supplier MilliporeSigma Supelco Supplier No. 6119350MG
Add to Cart
Add to Cart

Specifications

CAS 125769-67-7
Product Line Selectophore
Linear Formula C26H44N2S4
MDL Number MFCD00214160
Melting Point 91°C to 92°C
Synonym o-XBDiBDTC; o-Xylylenebis(N,N-diisobutyldithiocarbamate)
Type Organic Standard
Quantity 50 mg
Formula Weight 512.90
Grade Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.