Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Cyclohexanone, For ion-selective electrodes, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111019324 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9324 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9324 Supplier MilliporeSigma Supelco Supplier No. 29135100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclohexanone is an industrially important reagent. It is also a major intermediate in the synthesis of nylon.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00001625

Specifications

Melting Point -47°C
CAS 108-94-1
Density 0.947 g/mL (at 25°C)
Quantity 100 mL
Flash Point 44°C - Closed Cup
Ignition Point 788°F
Product Line Selectophore
Linear Formula C6H10(=O)
Vapor Pressure 3.4 mmHg (20°C)
Boiling Point 155°C
Formula Weight 98.14
Percent Purity ≥99.5% (GC); ≥99.5%
Show More Show Less
RTECSNumber GW1050000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.