Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Dioctyl phenylphosphonate, MilliporeSigma™ Supelco™

Catalog No. 111013356 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3356 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3356 Supplier MilliporeSigma Supelco Supplier No. 1258410ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1754-47-8
Molecular Weight (g/mol) 382.52
MDL Number MFCD00009561
Synonym DOPP

Specifications

Density 0.967 g/mL (at 25°C)
Boiling Point 207°C (4 mmHg)
Flash Point 113°C
Linear Formula C6H5 P(O)[O(CH2)7 CH3]2
Quantity 10 mL
Formula Weight 382.52
Grade Selectophore
Physical Form Liquid
Product Line Selectophore
RTECSNumber TA0379000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.