Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Dodecyl 2-nitrophenyl ether, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111015635 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5635 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5635 Supplier MilliporeSigma Supelco Supplier No. 442245ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dodecyl 2-nitrophenyl ether is an nitro-aromatic solvent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00024221
Synonym 1-Dodecyloxy-2-nitrobenzene; 2-Nitrophenyl dodecyl ether; ETH 217

Specifications

Density 0.999 g/mL (at 20°C)
CAS 83027-71-8
Boiling Point 218°C/1.5 mmHg
Flash Point 109°C
Linear Formula C18H29 NO3
Quantity 5 mL
Formula Weight 307.43
Percent Purity ≥99.0% (GC); ≥99.0%
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.