Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ hemi-Calcium bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate, ≥97.0%, MilliporeSigma™ Supelco™

Catalog No. 111014155 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4155 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4155 Supplier MilliporeSigma Supelco Supplier No. 151801G
Add to Cart
Add to Cart

Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt (hemi-Calcium bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate) is an ionophore of calcium.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00043425
Synonym Bis[4-(1,1,3,3-tetramethylbutyl)phenyl] phosphate calcium salt

Specifications

CAS 40835-97-0
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C56H84 CaO8P2
Quantity 1 g
Solubility Information THF: 2 mg/10 mL, clear, colorless
Formula Weight 987.29
Percent Purity ≥97.0% (HPLC); ≥97.0%
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.