Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Hydrogen ionophore I, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111012329 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2329 100 mg
11-101-2330 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2329 Supplier MilliporeSigma Supelco Supplier No. 95292100MG
Add to Cart
Add to Cart

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00008971
Synonym Tridodecylamine; Proton ionophore I; Hydrogen ionophore I

Specifications

Density 0.823 g/mL (at 25°C)
CAS 102-87-4
Boiling Point 220°C to 228°C/0.03 mmHg
Flash Point 113°C
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula [CH3(CH2)11]3 N
Quantity 100 mg
Vapor Density >1 (vs. air)
Formula Weight 521.99
Grade Selectophore
Product Line Selectophore
Show More Show Less

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.