Learn More
Selectophore™ Hydrogen ionophore I, Function Tested, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Density | 0.823 g/mL (at 25°C) |
| CAS | 102-87-4 |
| Boiling Point | 220°C to 228°C/0.03 mmHg |
| Flash Point | 113°C |
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| Linear Formula | [CH3(CH2)11]3 N |
| Quantity | 500 mg |
| Molecular Weight (g/mol) | 521.99 |
| Vapor Density | >1 (vs. air) |
| Formula Weight | 521.99 |
| Show More |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.