Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Lithium Ionophore III, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111016015 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
Packaging:
Bottomless Glass Bottle with Fused Cone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-6015 50 mg Bottomless Glass Bottle with Fused Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6015 Supplier MilliporeSigma Supelco Supplier No. 6255850MG

Lithium ionophore used in liquid membrane electrodes for the determination of Li concentration in blood

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99281-50-2
Molecular Formula C28H50N2O2
Molecular Weight (g/mol) 446.72
MDL Number MFCD00077936
InChI Key HEMVYQNWBJZPTO-BDBAHPOVSA-N
Synonym ETH 1810; N,N-Dicyclohexyl-N',N'-diisobutyl-cis-cyclohexane-1,2-dicarboxamide
IUPAC Name (1S,2R)-N1,N1-bis(butan-2-yl)-N2,N2-dicyclohexylcyclohexane-1,2-dicarboxamide
SMILES CCC(C)N(C(C)CC)C(=O)[C@H]1CCCC[C@H]1C(=O)N(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 110°C to 112°C
Packaging Bottomless Glass Bottle with Fused Cone
Linear Formula C28H50N2O2
Quantity 50 mg
Formula Weight 446.71
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.