Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Magnesium Ionophore VI, Function Tested, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013202 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
Packaging:
Bottomless Glass Bottle with Fused Cone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-101-3202 10 mg Bottomless Glass Bottle with Fused Cone
11-101-3201 100 mg Bottomless Glass Bottle with Fused Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3202 Supplier MilliporeSigma Supelco Supplier No. 6311210MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lipophilic magnesium-selective ionophore (log PTLC=8.5) with sufficient selectivity for Mg2+ against the main interfering cations for serum measurments

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151058-38-7
Molecular Formula C63H96N6O6
Molecular Weight (g/mol) 1033.50
MDL Number MFCD01867417
InChI Key ODMHCQNFQKBJOO-UHFFFAOYSA-N
Synonym 1,3,5-Tris[10-(1-adamantyl)-7,9-dioxo-6,10-diazaundecyl]benzene; ETH 5506
IUPAC Name N'-(adamantan-1-yl)-N-{5-[3,5-bis(5-{2-[(adamantan-1-yl)(methyl)carbamoyl]acetamido}pentyl)phenyl]pentyl}-N'-methylpropanediamide
SMILES CN(C(=O)CC(=O)NCCCCCC1=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=CC(CCCCCNC(=O)CC(=O)N(C)C23CC4CC(CC(C4)C2)C3)=C1)C12CC3CC(CC(C3)C1)C2

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
Linear Formula C63H96N6O6
Quantity 10 mg
Formula Weight 1033.47
Percent Purity ≥99.0% (HPLC)
Grade Selectophore
Physical Form Solid
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.