Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Nitrate Ionophore VI, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111012732 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2732 10 mg
11-101-2731 100 mg
11-101-2730 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-2732 Supplier MilliporeSigma Supelco Supplier No. 0729510MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

New ionophore for online monitoring of nitrate obtains superior characteristics over all existing nitrate ionophores.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD16652520
Synonym 9-Hexadecyl-1,7,11,17-tetraoxa-2,6,12,16-tetraazacycloeicosane

Specifications

CAS 1196157-85-3
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C28H60N4O4
Quantity 10 mg
Formula Weight 516.80
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.