Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Nitrite ionophore I, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111012921 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2921 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2921 Supplier MilliporeSigma Supelco Supplier No. 7259010MG
Add to Cart
Add to Cart

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00213827
Synonym Cyanoaqua-cobyrinic acid heptakis(2-phenylethyl ester)

Specifications

CAS 130549-56-3
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C102H117 ClCoN5O19
Quantity 10 mg
Formula Weight 1811.43
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.