Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Perfluoroperhydrophenanthrene, MilliporeSigma™ Supelco™

Catalog No. 111013879 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3879 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3879 Supplier MilliporeSigma Supelco Supplier No. 5691910ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Description Solvent for preparation of membranes
CAS 306-91-2
Product Line Selectophore
Density 2.03 g/mL (at 20°C)
Linear Formula C14F24
MDL Number MFCD00042596
Boiling Point 212°C to 218°C
For Use With (Equipment) Ion Selective Electrode
Type Organic Standard
Quantity 10 mL
Formula Weight 624.11
Show More Show Less

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.