Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Poly(vinyl chloride) carboxylated, 1.8 carboxyl basis, 1.8% carboxyl basis, MilliporeSigma™ Supelco™

Catalog No. 111018330 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8330 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8330 Supplier MilliporeSigma Supelco Supplier No. 8139550G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00084460
Synonym Poly(vinyl chloride-co-acrylic acid)

Specifications

CAS 25702-80-1
Linear Formula (C3H4O2 · C2H3 Cl)x
Quantity 50 g
Percent Purity 1.8% carboxyl basis
Grade Selectophore
Physical Form Powder
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.