Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Potassium tetrakis(4-chlorophenyl)borate, ≥98.0%, MilliporeSigma™ Supelco™

Catalog No. 111013145 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3145 5 g
11-101-3143 100 mg
11-101-3144 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-3145 Supplier MilliporeSigma Supelco Supplier No. 605915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00043105
Synonym Tetrakis(4-chlorophenyl)boron potassium

Specifications

CAS 14680-77-4
Packaging Glass Bottle
Linear Formula (ClC6H4)4 BK
Quantity 5 g
Formula Weight 496.11
Percent Purity ≥98.0% (AT); ≥98.0%
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.