Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, ≥98.0% (based on Na, AAS), MilliporeSigma™ Supelco™

Catalog No. 111015551 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5551 10 mg
11-101-5553 50 mg
11-101-5552 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-5551 Supplier MilliporeSigma Supelco Supplier No. 7201710MG
Add to Cart
Add to Cart

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic ion which is insoluble in water. It is also strong against acids and oxidants. It can be prepared using boron trifluoride and 3,5-bis(trifluoromethyl)phenylmagnesium iodide as major components.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00043323
Synonym NaBARF; Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron sodium

Specifications

CAS 79060-88-1
Quantity 10 mg
Linear Formula C32H12 BF24 Na
Formula Weight 886.20
Percent Purity ≥98.0% (based on Na, AAS)
Grade Selectophore
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.