Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Tetradodecylammonium tetrakis(4-chlorophenyl)borate, MilliporeSigma™ Supelco™

Catalog No. 111012275 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2275 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2275 Supplier MilliporeSigma Supelco Supplier No. 872551G
Add to Cart
Add to Cart

ETH 500 is a lipophilic electrolyte mostly used as an anionic additive.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00145372
Synonym ETH 500; Tetrakis(4-chlorophenyl)borate tetradodecylammonium salt

Specifications

Melting Point 79°C to 80°C
CAS 100581-42-8
Linear Formula C72H116 BCl4 N
Quantity 1 g
Formula Weight 1148.32
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.