Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Tetraoctylammonium nitrate, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013960 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3960 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3960 Supplier MilliporeSigma Supelco Supplier No. 87983500MG
Add to Cart
Add to Cart

Tetraoctylammonium nitrate (TOAN) is an ionophore or recognition element mostly used in potentiometric sensors.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00525561

Specifications

Melting Point 112°C to 114°C
CAS 33734-52-0
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula [CH3(CH2)6 CH2]4 N(NO3)
Quantity 500 mg
Formula Weight 528.89
Percent Purity ≥99.0% (NT); ≥99.0%
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.