Learn More
Medchemexpress LLC Selegiline hydrochloride | 14611-52-0 | 556605 | 99.94% | C13H18ClN | 250 MG
SDP
Catalog No. 5000357343
Encompass_Preferred
Selegiline hydrochloride, also known as Deprenyl hydrochloride, is a potent, selective, and irreversible inhibitor of MAO-B, with an IC50 of 51 nM. It exhibits 450-fold selectivity for MAO-B over MAO-A. This compound is used for research related to Parkinson's disease, Alzheimer's disease, and major depressive disorder.
  • Potent, selective, and irreversible MAO-B inhibitor
  • Exhibits high selectivity for MAO-B over MAO-A
  • Supports research into Parkinson's disease
  • Supports research into Alzheimer's disease
  • Supports research into major depressive disorder
  • Inhibits recombinant human MAO-B in a concentration-dependent manner
  • Increases efflux of norepinephrine, dopamine, and serotonin
Catalog No. 50-003-57343 Supplier Medchemexpress LLC Supplier No. 250MGHY14199
Add to Cart
Add to Cart
Selegiline hydrochloride, also known as Deprenyl hydrochloride, is a potent, selective, and irreversible inhibitor of MAO-B, with an IC50 of 51 nM. It exhibits 450-fold selectivity for MAO-B over MAO-A. This compound is used for research related to Parkinson's disease, Alzheimer's disease, and major depressive disorder.
  • Potent, selective, and irreversible MAO-B inhibitor
  • Exhibits high selectivity for MAO-B over MAO-A
  • Supports research into Parkinson's disease
  • Supports research into Alzheimer's disease
  • Supports research into major depressive disorder
  • Inhibits recombinant human MAO-B in a concentration-dependent manner
  • Increases efflux of norepinephrine, dopamine, and serotonin