Learn More
Medchemexpress LLC Selegiline hydrochloride | 14611-52-0 | MFCD00069299 | 99.94% | C13H18ClN | 1 ML
SDP
Catalog No. 5000354373
Encompass_Preferred
Selegiline hydrochloride (Deprenyl hydrochloride) is a potent, selective and irreversible inhibitor of MAO-B, with an IC50 of 51 nM. It exhibits 450-fold selectivity for MAO-B over MAO-A (IC50=23 μM). Selegiline hydrochloride can be used for the research of Parkinson's disease, Alzheimer's disease and major depressive disorder.
  • Potent, selective, and irreversible inhibitor of MAO-B
  • Exhibits 450-fold selectivity for MAO-B over MAO-A
  • Increases efflux of norepinephrine, dopamine, and serotonin from rat hypothalamus
  • Inhibits efflux of dihydroxyphenylacetic acid and 5-hydroxyindoleacetic acid
  • Reduces cocaine self-administration in mice
  • Reduces DOPAC and 5-HIAA concentrations in the frontal cortex
Catalog No. 50-003-54373 Supplier Medchemexpress LLC Supplier No. HY1419910MM/1ML
Selegiline hydrochloride (Deprenyl hydrochloride) is a potent, selective and irreversible inhibitor of MAO-B, with an IC50 of 51 nM. It exhibits 450-fold selectivity for MAO-B over MAO-A (IC50=23 μM). Selegiline hydrochloride can be used for the research of Parkinson's disease, Alzheimer's disease and major depressive disorder.
  • Potent, selective, and irreversible inhibitor of MAO-B
  • Exhibits 450-fold selectivity for MAO-B over MAO-A
  • Increases efflux of norepinephrine, dopamine, and serotonin from rat hypothalamus
  • Inhibits efflux of dihydroxyphenylacetic acid and 5-hydroxyindoleacetic acid
  • Reduces cocaine self-administration in mice
  • Reduces DOPAC and 5-HIAA concentrations in the frontal cortex