Learn More
NEOGEN Selenite Broth, 500 g
SDP
Catalog No. 502015648
Encompass_Preferred
Selenite Broth is used for the selective enrichment of Salmonella spp. in a laboratory setting. Selenite Broth is not intended for use in the diagnosis of disease or other conditions in humans. Selenite Broth was originated by Leifson, while observing good recovery of Salmonella spp. and reduced growth of fecal coliforms. Salmonella organisms are also injured in food-processing procedures, including exposure to low temperatures, sub-marginal heat, drying, radiation, preservatives or sanitizers. Selenite Broth conforms with the American Public Health Association (APHA). Many modifications of Selenite Broth exist, including Selenite Cystine Broth, from the original formula described as Selenite F Broth by Leifson.For more information visit https://www.neogen.com/solutions/Microbiology/selenite-broth
Catalog No. 50-201-5648 Supplier NEOGEN Supplier No. 700003476
Add to Cart
Add to Cart
Selenite Broth is used for the selective enrichment of Salmonella spp. in a laboratory setting. Selenite Broth is not intended for use in the diagnosis of disease or other conditions in humans. Selenite Broth was originated by Leifson, while observing good recovery of Salmonella spp. and reduced growth of fecal coliforms. Salmonella organisms are also injured in food-processing procedures, including exposure to low temperatures, sub-marginal heat, drying, radiation, preservatives or sanitizers. Selenite Broth conforms with the American Public Health Association (APHA). Many modifications of Selenite Broth exist, including Selenite Cystine Broth, from the original formula described as Selenite F Broth by Leifson.For more information visit https://www.neogen.com/solutions/Microbiology/selenite-broth