Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Selenite Broth

Catalog No. R064502
Change view
Click to view available options
Quantity:
20/Pk.
Format:
Tube
Media Type:
Selenite Broth
Manufacturing Origin:
United States
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
R064502 20/Pk. Tube Selenite Broth United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R064502 Supplier Thermo Scientific™ Supplier No. R064502
This item is not returnable. View return policy

Reduce media preparation time and simultaneously enrich and isolate Salmonella spp. using convenient, pre-filled, ready-to-use tubes.

Reduce media preparation time and simultaneously enrich and isolate Salmonella spp. using convenient, pre-filled, ready-to-use Thermo Scientific™ Selenite Broth.

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Selenite Broth.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear liquid, faint to very light amber
Content And Storage 2°C to 8°C
Format Tube
Manufacturing Origin United States
Media Type Selenite Broth
Product Type Microbiology Media
Fill Volume 9 mL
Packaging Type Glass tube with black screw cap
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.