Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Selenite Cystine Broth
SDP

Catalog No. 23001770 Shop All Hardy Diagnostics Products
This item is not returnable. View return policy
Catalog No. 23-001-770 Supplier Hardy Diagnostics™ Supplier No. K69
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selenite Cystine Broth is recommended for the selective enrichment of Salmonella spp. in clinical and non-clinical specimens.

  • Useful in detecting Salmonella when low numbers of organisms are present in stool
  • Recommended for use in detecting Salmonella in food and water

Specifications

Appearance Clear
Broth Type Selenite Cystine Broth
Certifications/Compliance ISO 13485
Color Light Orange
Content And Storage 2°C to 8°C
Additive Sodium Selenite, L-Cystine
Detects Salmonella spp.
Format Tube
Manufacturing Origin United States
Media Type Broth
Product Type Broth Tube
Target Organism Class Gram-negative Rods
Test Type Environmental Test
Fill Volume 10 mL
Formulation Sodium Phosphate - 10.0 gm, Tryptone - 5.0 gm, Lactose - 4.0 gm, Sodium Selenite - 4.0 gm, L-Cystine - 0.01 gm
For Use With (Application) Breweries, Wineries and Distilleries, Cannabis QA Testing, Clinical, Cosmetics, Environmental, Food Safety
Incubation Conditions Aerobically at 35°C
Incubation Time 18 to 24 hr.
Material Polycarbonate
No. per Pack 20
Packaging Type Box
Quantity 20 x 10 mL
Dimensions (Dia. x H) 16 x 125 mm
Height (Metric) 125 mm
Diameter (Metric) 16 mm
Protein Source Casein
Source Casein
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.