Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALADDIN SCIENTIFIC CORPORATION Selenium, 25g

Catalog No. 5000082501
Encompass_Preferred

Selenium 99 9% metals basis 200 meshCas-7782-49-2

Catalog No. 50-000-82501 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. S10519325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selenium 99 9% metals basis 200 meshCas-7782-49-2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.