Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium +6 Assurance™ Grade Single Speciation Standard, 125 mL in H2O, SPEX CertiPrep™
SDP

Catalog No. SPECSE6 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPECSE6 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPECSE6 Supplier SPEX CertiPrep Supplier No. SPECSE6
Add to Cart
Add to Cart

Includes

Certificate of Analysis

Selenium +6 Assurance™ Grade Single Speciation Standard for sample verification and method validation, 125 mL in H2O, supplied with a comprehensive Certificate of Analysis

  • Selenium +3 Speciation Standard
  • 125 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Chemical Identifiers

CAS 7782-49-2 , 7732-18-5
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
IUPAC Name selenium
SMILES [Se]

Specifications

Description Speciation +6 Speciation Standard
Packaging EA
CAS Min % 0.1%, 0.00%
CAS Max % 99.9%, 0.00%
Analyte or Component Names Selenium +6
Concentration 1000 μg/mL
Technique ICP-MS
Certifications/Compliance ISO/IEC 17025, ISO 17034, ISO 9001
UN Number 12352402
Type Certified Reference Material
Element Selenium +6
Quantity 1 Ea.
Show More Show Less
ShelfLife 1 year
Recommended Storage Ambient Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.