Learn More
Cusabio Technology LLC selenium binding protein 1
Catalog No. 5000048921
Encompass_Preferred
The Human Selenium Binding Protein 1 (SELENBP1) ELISA Kit is designed for the quantitative detection of SELENBP1 in serum plasma and tissue homogenates using a sandwich ELISA method This kit detects SELENBP1 within a broad concentration range of 125 pg/mL to 8000 pg/mL with a sensitivity threshold of 31 25 pg/mL ensuring precise measurement across diverse biological samples The assay requires a sample volume of 50-100 L and delivers results within 1 to 5 hours of incubation time By employing a 450 nm wavelength for absorbance measurement it provides reliable and reproducible data for research applications Suitable for both routine analysis and specialized studies this kit offers a robust tool for investigating SELENBP1 levels in physiological or pathological contexts
Catalog No. 50-000-48921 Supplier Cusabio Technology LLC Supplier No. CSBE13947H
The Human Selenium Binding Protein 1 (SELENBP1) ELISA Kit is designed for the quantitative detection of SELENBP1 in serum plasma and tissue homogenates using a sandwich ELISA method This kit detects SELENBP1 within a broad concentration range of 125 pg/mL to 8000 pg/mL with a sensitivity threshold of 31 25 pg/mL ensuring precise measurement across diverse biological samples The assay requires a sample volume of 50-100 L and delivers results within 1 to 5 hours of incubation time By employing a 450 nm wavelength for absorbance measurement it provides reliable and reproducible data for research applications Suitable for both routine analysis and specialized studies this kit offers a robust tool for investigating SELENBP1 levels in physiological or pathological contexts