Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium, Black 99+, MilliporeSigma™

Catalog No. p-4939074 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1077140050 50 g Plastic bottle
M1077140250 250 g Plastic bottle
M1077141000 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1077140050 Supplier MilliporeSigma Supplier No. 1077140050
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7782-49-2
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]

Specifications

Physical Form Solid
Quantity 50 g
Formula Weight 78.96
Odor Odorless
Packaging Plastic bottle
Appearance Dark gray solid
Chemical Name or Material Selenium
Hazard Category H331: Toxic if inhaled.
H301: Toxic if swallowed.
H373: May cause damage to organs through prolonged or repeated exposure.
H413: May cause long lasting harmful effects to aquatic life.
EINECSNumber 231-957-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.