Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Selenium Nanoparticles 150nm 0.15 Wt% Aqueous Dispersion

Catalog No. 9195271ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-952-71ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-952-71ML Supplier Sigma Aldrich Supplier No. 9195271ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Selenium nanoparticles (SeNPs) are used in a wide variety of applications such as antimicrobial coatings, nanotherapeutics, diagnostics, and medical devices. These nanoparticles have been shown to play an important role in protecting against oxidative stress and have demonstrated low toxicity and bioavailability.

Specifications

CAS 7782-49-2
Quantity 1 mL
Molecular Formula Se
Linear Formula Se
Synonym SeNP
Molecular Weight (g/mol) 78.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.