Learn More
Strem, An Ascensus Company CAS# 7782-49-2. 100g. Selenium powder (99.5%). MFCD00134090
SDP
Catalog No. 5090115122
Encompass
CAS# 7782-49-2. 100g. Selenium powder (99.5%). MFCD00134090. Molecular Weight: 78.96. Molecular Formula: Se. Color/form: -200 mesh Strem# 93-3416. http://www.strem.com/catalog/v/93-3416/
Catalog No. 50-901-15122 Supplier Strem, An Ascensus Company Supplier No. 933416100G
CAS# 7782-49-2. 100g. Selenium powder (99.5%). MFCD00134090. Molecular Weight: 78.96. Molecular Formula: Se. Color/form: -200 mesh Strem# 93-3416. http://www.strem.com/catalog/v/93-3416/