Learn More
Medchemexpress LLC Selenocystamine dih 25g
SDP
Catalog No. 5000204640
Encompass_Preferred
Selenocystamine dihydrochloride is a selenocysteine derivative that can be used in the synthesis of other active compounds Selenocystamine dihydrochloride can also induce the aggregation of amphiphilic p-sulfonatocalixarene to form supramolecular nanoparticles[1][2][3]
Catalog No. 50-002-04640 Supplier Medchemexpress LLC Supplier No. 25GHYW105804
Selenocystamine dihydrochloride is a selenocysteine derivative that can be used in the synthesis of other active compounds Selenocystamine dihydrochloride can also induce the aggregation of amphiphilic p-sulfonatocalixarene to form supramolecular nanoparticles[1][2][3]