Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Selenoethionine | 2578-27-0 | 210.13 g·mol⁻¹ | C6H13NO2Se | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW58582510MG

Encompass_Preferred

Selenoethionine is a selenium-containing amino acid supplied for laboratory research use. It serves as a selenium source for incorporation into proteins, metabolism studies, and enzymatic assays. Intended for research use only; consult the certificate of analysis for analytical and storage details.

  • Selenium-containing amino acid for research use.
  • Chemical formula: C6H13NO2Se.
  • Molecular weight: 210.13 g·mol⁻¹.
  • CAS number: 2578-27-0.
  • Supplied as a 10 mg vial.
  • Certificate of analysis available for analytical data.
  • Store under conditions specified in the certificate of analysis.

Catalog No. 50-002-84557


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.